missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine Hydrobromide 98.0+%, TCI America™

Catalog No. D46671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46671G 1 g
D46675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46671G Supplier TCI America Supplier No. D46671G

Chemical Identifiers

CAS 6274-12-0
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD00012498
InChI Key AATGHKSFEUVOPF-UHFFFAOYSA-N
Synonym diethylamine hydrobromide, ethanamine, n-ethyl-, hydrobromide, diethylamine hbr, acmc-1b9lr, n-ethylethanamine hydrobromide
PubChem CID 10986453
IUPAC Name diethylazanium bromide
SMILES [Br-].CC[NH2+]CC

Specifications

Melting Point 219°C
Color White
Quantity 1 g
Formula Weight 154.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylamine Hydrobromide