missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 99+%, Thermo Scientific Chemicals

Catalog No. AAA11716AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11716AP 500 mL
AAA11716AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA11716AP Supplier Thermo Scientific Chemicals Supplier No. A11716AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).

Solubility
Miscible with water, alcohol, ether, chloroform and carbon terachloride.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
SMILES CCNCC

Specifications

Melting Point °C to 50°C
Density 0.706
Boiling Point 54°C to 55°C
Flash Point -28°C (-18°F)
Assay Percent Range ≥99%
Refractive Index 1.385
Quantity 500 mL
Beilstein 605268
Sensitivity Air Sensitive
Solubility Information Miscible with water,alcohol,ether,chloroform and carbon terachloride.
IUPAC Name N-ethylethanamine
Formula Weight 73.14
Physical Form Liquid
Chemical Name or Material Diethylamine
Show More Show Less
Hazard Category H225-H302+H332-H311-H314-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-716-3
RTECSNumber HZ8750000
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only