Learn More
Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals
$98.03 - $98.03
Chemical Identifiers
| CAS | 109-89-7 |
|---|---|
| Molecular Formula | C4H11N |
| Molecular Weight (g/mol) | 73.14 |
| MDL Number | MFCD00009032 |
| InChI Key | HPNMFZURTQLUMO-UHFFFAOYSA-N |
| Synonym | diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine |
| PubChem CID | 8021 |
| ChEBI | CHEBI:85259 |
| IUPAC Name | N-ethylethanamine |
| SMILES | CCNCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC458492500
|
Thermo Scientific Chemicals
458492500 |
250 mL |
Each for $98.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109-89-7 | |
| 73.14 | |
| HPNMFZURTQLUMO-UHFFFAOYSA-N | |
| 8021 | |
| N-ethylethanamine |
| C4H11N | |
| MFCD00009032 | |
| diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
| CHEBI:85259 | |
| CCNCC |
Specifications
| 109-89-7 | |
| 0.7100g/mL | |
| −23°C | |
| C4H11N | |
| (C2H5)2NH | |
| 250 mL | |
| diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
| HPNMFZURTQLUMO-UHFFFAOYSA-N | |
| N-ethylethanamine | |
| 8021 | |
| CHEBI:85259 | |
| 99.5% | |
| Diethylamine |
| -50.0°C | |
| 55.0°C | |
| 99.4% min. (GC) | |
| 1.3840 to 1.3860 | |
| MFCD00009032 | |
| 0.71 | |
| Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents | |
| CCNCC | |
| 73.14 | |
| 0.34 mPa.s (25°C) | |
| 73.13 | |
| Analysis |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 203-716-3
RUO – Research Use Only