missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog No. AC458492500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458492500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458492500 Supplier Thermo Scientific Chemicals Supplier No. 458492500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
IUPAC Name N-ethylethanamine
SMILES CCNCC

Specifications

Melting Point -50.0°C
Density 0.7100g/mL
Boiling Point 55.0°C
Flash Point −23°C
Assay Percent Range 99.4% min. (GC)
Refractive Index 1.3840 to 1.3860
Linear Formula (C2H5)2NH
Quantity 250 mL
Specific Gravity 0.71
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents
Viscosity 0.34 mPa.s (25°C)
Formula Weight 73.13
Percent Purity 99.5%
Grade Analysis
Chemical Name or Material Diethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-716-3

RUO – Research Use Only