missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 0.10 Normal, Standardized, Reagents

Catalog No. 01335424
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335424 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335424 Supplier Labchem Inc Supplier No. CD2270004C

Diethylamine, 0.10 Normal, Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Diethylamine, 0.10 Normal, Standardized
CAS 7732-18-5, 109-89-7
Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.9969
Synonym N-Ethylethanamine, N,N-Diethylamine
Grade Reagent
Physical Form Liquid