missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Vinylphosphonate 98.0+%, TCI America™

Catalog No. D37285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37285G Supplier TCI America Supplier No. D37285G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 682-30-4
Molecular Formula C6H13O3P
Molecular Weight (g/mol) 164.14
MDL Number MFCD00009079
InChI Key DREPONDJUKIQLX-UHFFFAOYSA-N
Synonym diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g
PubChem CID 69629
IUPAC Name diethyl ethenylphosphonate
SMILES CCOP(=O)(OCC)C=C

Specifications

Color Colorless
Boiling Point 82°C
Quantity 5 g
Formula Weight 164.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Vinylphosphonate
RTECSNumber TA1150000
TSCA No
Recommended Storage Refrigerator