missing translation for 'onlineSavingsMsg'
Learn More

Diethyl vinylphosphonate, 97%

Catalog No. p-3735848
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152150100 10 g Glass bottle
AC152150250 25 g Glass bottle
AC152151000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC152150100 Supplier Thermo Scientific Chemicals Supplier No. 152150100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 682-30-4 | C6H13O3P | 164.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 682-30-4
Molecular Formula C6H13O3P
Molecular Weight (g/mol) 164.14
MDL Number MFCD00009079
InChI Key DREPONDJUKIQLX-UHFFFAOYSA-N
Synonym diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g
PubChem CID 69629
IUPAC Name 1-[ethenyl(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)C=C

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 202°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.43
Linear Formula (C2H5O)2P(O)CH=CH2
Quantity 10 g
Specific Gravity 1.06
Formula Weight 164.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl vinylphosphonate
Show More Show Less
EINECSNumber 211-663-2

RUO – Research Use Only