missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Trichloromethyl)phosphonate 96.0+%, TCI America™

Catalog No. D460725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D460725G 25 g
D46075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D460725G Supplier TCI America Supplier No. D460725G

Chemical Identifiers

CAS 866-23-9
Molecular Formula C5H10Cl3O3P
Molecular Weight (g/mol) 255.46
MDL Number MFCD00013666
InChI Key RVAQSYWDOSHWGP-UHFFFAOYSA-N
Synonym diethyl trichloromethylphosphonate, diethyl trichloromethyl phosphonate, phosphonic acid, trichloromethyl-, diethyl ester, methanephosphonic acid, trichloro-, diethyl ester, diethyltrichloromethylphosphonate, wln: gxggpo&o2&o2, 4-03-00-00262 beilstein handbook reference, trichloromethylphosphonic acid diethyl, diethoxyphosphino trichloromethyl-1-one
PubChem CID 70085
IUPAC Name diethyl (trichloromethyl)phosphonate
SMILES CCOP(=O)(OCC)C(Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 g
Formula Weight 255.46
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Trichloromethyl)phosphonate
RTECSNumber TA0988000
TSCA No
Recommended Storage Refrigerator