missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-4-Cyclohexene-1,2-dicarboxylate 97.0+%, TCI America™

Catalog No. C17045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17045G 5 g
C17041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17045G Supplier TCI America Supplier No. C17045G
Only null left

Chemical Identifiers

CAS 5048-50-0
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD06799457
InChI Key OWVOHDKOANTMJU-UHFFFAOYNA-N
Synonym trans-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester, Diethyl trans-1,2,3,6-Tetrahydrophthalate, trans-1,2,3,6-Tetrahydrophthalic Acid Diethyl Ester
PubChem CID 12358152
IUPAC Name 1,2-diethyl cyclohex-4-ene-1,2-dicarboxylate
SMILES CCOC(=O)C1CC=CCC1C(=O)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 226.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl trans-4-Cyclohexene-1,2-dicarboxylate
EINECSNumber (3)-2437
TSCA No