missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-3-Hexenedioate 98.0+%, TCI America™

Catalog No. B159210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B159210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B159210G Supplier TCI America Supplier No. B159210G

Chemical Identifiers

CAS 57042-08-7
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00191608
InChI Key YIDKJPKYCLMJQW-AATRIKPKSA-N
Synonym trans-2-Butene-1,4-dicarboxylic Acid Diethyl Ester, trans-3-Hexenedioic Acid Diethyl Ester, Diethyl trans-2-Butene-1,4-dicarboxylate
PubChem CID 6148048
IUPAC Name diethyl (E)-hex-3-enedioate
SMILES CCOC(=O)CC=CCC(=O)OCC

Specifications

Color Colorless
Quantity 10 g
Formula Weight 200.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl trans-3-Hexenedioate