missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl trans-1,4-Cyclohexanedicarboxylate 98.0+%, TCI America™
$255.38 - $762.40
Chemical Identifiers
| CAS | 19145-96-1 |
|---|---|
| Molecular Formula | C12H20O4 |
| Molecular Weight (g/mol) | 228.288 |
| InChI Key | KRJHRNUTLDTSKY-UHFFFAOYSA-N |
| Synonym | trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester |
| PubChem CID | 254555 |
| IUPAC Name | diethyl cyclohexane-1,4-dicarboxylate |
| SMILES | CCOC(=O)C1CCC(CC1)C(=O)OCC |
Chemical Identifiers
| 19145-96-1 | |
| 228.288 | |
| trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester | |
| diethyl cyclohexane-1,4-dicarboxylate |
| C12H20O4 | |
| KRJHRNUTLDTSKY-UHFFFAOYSA-N | |
| 254555 | |
| CCOC(=O)C1CCC(CC1)C(=O)OCC |
Specifications
| 19145-96-1 | |
| White-Yellow | |
| 5 g | |
| KRJHRNUTLDTSKY-UHFFFAOYSA-N | |
| diethyl cyclohexane-1,4-dicarboxylate | |
| 254555 | |
| ≥98.0% (GC) | |
| Diethyl trans-1,4-Cyclohexanedicarboxylate |
| 45°C | |
| C12H20O4 | |
| trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester | |
| CCOC(=O)C1CCC(CC1)C(=O)OCC | |
| 228.288 | |
| 228.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No