missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-1,4-Cyclohexanedicarboxylate 98.0+%, TCI America™

Catalog No. D36395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36395G 5 g
D363925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36395G Supplier TCI America Supplier No. D36395G
Only null left

Chemical Identifiers

CAS 19145-96-1
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.288
InChI Key KRJHRNUTLDTSKY-UHFFFAOYSA-N
Synonym trans-1,4-Cyclohexanedicarboxylic Acid Diethyl Ester
PubChem CID 254555
IUPAC Name diethyl cyclohexane-1,4-dicarboxylate
SMILES CCOC(=O)C1CCC(CC1)C(=O)OCC

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl trans-1,4-Cyclohexanedicarboxylate