missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-1,2-Cyclopropanedicarboxylate 96.0+%, TCI America™

Catalog No. p-7128435
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47431G 1 g
D47435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47431G Supplier TCI America Supplier No. D47431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3999-55-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD00198002
InChI Key SXLDHZFJMXLFJU-RNFRBKRXSA-N
Synonym diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane
PubChem CID 11715303
IUPAC Name diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
SMILES CCOC(=O)C1CC1C(=O)OCC

Specifications

Color Yellow
Boiling Point 71°C
Quantity 1 g
Formula Weight 186.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl trans-1,2-Cyclopropanedicarboxylate