missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate 96.0+%, TCI America™
$147.25 - $512.62
Chemical Identifiers
| CAS | 71885-51-3 |
|---|---|
| Molecular Formula | C10H21O5P |
| Molecular Weight (g/mol) | 252.247 |
| MDL Number | MFCD09263515 |
| InChI Key | RUPVGTAIRUGQIQ-UHFFFAOYSA-N |
| Synonym | [(Tetrahydropyran-2-yloxy)methyl]phosphonic Acid Diethyl Ester |
| PubChem CID | 11230495 |
| IUPAC Name | 2-(diethoxyphosphorylmethoxy)oxane |
| SMILES | CCOP(=O)(COC1CCCCO1)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4611200MG
|
TCI America
D4611200MG |
200 mg |
Each for $147.25
|
|
|||||
|
D46111G
|
TCI America
D46111G |
1 g |
Each for $512.62
|
|
|||||
Chemical Identifiers
| 71885-51-3 | |
| 252.247 | |
| RUPVGTAIRUGQIQ-UHFFFAOYSA-N | |
| 11230495 | |
| CCOP(=O)(COC1CCCCO1)OCC |
| C10H21O5P | |
| MFCD09263515 | |
| [(Tetrahydropyran-2-yloxy)methyl]phosphonic Acid Diethyl Ester | |
| 2-(diethoxyphosphorylmethoxy)oxane |
Specifications
| 71885-51-3 | |
| 95°C | |
| MFCD09263515 | |
| [(Tetrahydropyran-2-yloxy)methyl]phosphonic Acid Diethyl Ester | |
| CCOP(=O)(COC1CCCCO1)OCC | |
| 252.247 | |
| 252.25 | |
| Liquid |
| Yellow | |
| C10H21O5P | |
| 200 mg | |
| RUPVGTAIRUGQIQ-UHFFFAOYSA-N | |
| 2-(diethoxyphosphorylmethoxy)oxane | |
| 11230495 | |
| ≥96.0% (GC) | |
| Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate |
Safety and Handling
TSCA : No