missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 95°C |
| Quantity | 200 mg |
| Formula Weight | 252.25 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate |
Safety and Handling
| TSCA | No |