missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tert-butylmalonate, 96%

Catalog No. p-7045019
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0913114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0913114 Supplier Thermo Scientific Chemicals Supplier No. L0913114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 759-24-0 | C11H20O4 | 216.28 g/mol

Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 759-24-0
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00009152
InChI Key RJNICNBRGVKNSR-UHFFFAOYSA-N
Synonym diethyl tert-butylmalonate, diethyl t-butylmalonate, diethyl 1,1-dimethylethyl malonate, diethyl 2-tert-butyl malonate, propanedioic acid, 1,1-dimethylethyl-, diethyl ester, diethyltert-butylmalonate, diethyl 2-tert-butylmalonate #, tert-butylmalonic acid diethyl ester, 1,3-diethyl 2-tert-butylpropanedioate
PubChem CID 69798
IUPAC Name diethyl 2-tert-butylpropanedioate
SMILES CCOC(=O)C(C(=O)OCC)C(C)(C)C

Specifications

Density 0.978
Boiling Point 102°C to 104°C (11 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.425
Quantity 25 g
Beilstein 1781709
Solubility Information Immiscible with water.
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Diethyl tert-butylmalonate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 212-067-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only