missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Sulfite 98.0+%, TCI America™
$93.21 - $775.44
Chemical Identifiers
| CAS | 623-81-4 |
|---|---|
| Molecular Formula | C4H10O3S |
| Molecular Weight (g/mol) | 138.18 |
| MDL Number | MFCD00009098 |
| InChI Key | NVJBFARDFTXOTO-UHFFFAOYSA-N |
| Synonym | sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester |
| PubChem CID | 12197 |
| IUPAC Name | diethyl sulfite |
| SMILES | CCOS(=O)OCC |
Chemical Identifiers
| 623-81-4 | |
| 138.18 | |
| NVJBFARDFTXOTO-UHFFFAOYSA-N | |
| 12197 | |
| CCOS(=O)OCC |
| C4H10O3S | |
| MFCD00009098 | |
| sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester | |
| diethyl sulfite |
Specifications
| 623-81-4 | |
| 157°C | |
| MFCD00009098 | |
| 1993 | |
| NVJBFARDFTXOTO-UHFFFAOYSA-N | |
| diethyl sulfite | |
| 12197 | |
| ≥98.0% (GC) | |
| Diethyl Sulfite |
| Colorless | |
| C4H10O3S | |
| 25 g | |
| sulfurous acid, diethyl ester, ethyl sulfite, diethyl sulphite, ethyl sulfite, et2so3, diethylsulfite, ethoxysulfinyl ethoxy, diethylsulfit, sulfurous acid diethyl, diethyl sulfite, sulfurous acid diethyl ester | |
| CCOS(=O)OCC | |
| 138.18 | |
| 138.18 | |
| Liquid |
Safety and Handling
RTECSNumber : WT3511000
TSCA : Yes