missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Sulfate 98.0+%, TCI America™

Catalog No. D052525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D052525ML 25 mL
D0525500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D052525ML Supplier TCI America Supplier No. D052525ML

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC

Specifications

Melting Point -25°C
Color Colorless
Boiling Point 208°C
Quantity 25 mL
UN Number 1594
Formula Weight 154.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Sulfate
EINECSNumber (2)-1673
RTECSNumber WS7875000
TSCA Yes