missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfate, 99%

CAS: 64-67-5 | C4H10O4S | 154.18 g/mol

$141.47 - $141.47

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114560010
SDS View Documents
Thermo Scientific Chemicals
114560010
1 L Glass bottle
Each for $141.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Chemical Identifiers

64-67-5
154.18
DENRZWYUOJLTMF-UHFFFAOYSA-N
6163
diethyl sulfate
C4H10O4S
MFCD00009099
sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
CHEBI:34699
CCOS(=O)(=O)OCC

Specifications

64-67-5
1.1795g/mL
104°C
98.5% min. (GC)
C4H10O4S
(C2H5O)2SO2
1 L
01,253
15,3149
Solubility in water: 5g/L (20°C). Other solubilities: decomposes in water,soluble in alcohol and ether
CCOS(=O)(=O)OCC
154.18
1.79 mPa.s (20°C)
154.18
Liquid
-24.0°C
208.0°C
Authentic
Glass bottle
1.3990 to 1.4010
MFCD00009099
01,327
1.1795
sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
DENRZWYUOJLTMF-UHFFFAOYSA-N
diethyl sulfate
6163
CHEBI:34699
99%
Diethyl sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause genetic defects.
May cause cancer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 200-589-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only