missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Succinate 99.0+%, TCI America™

Catalog No. p-7135809
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
S010325G 25 g
S0103500G 500 g
2 options

Catalog No. S010325G

Supplier: TCI America S010325G

Only null left

Chemical Identifiers

CAS 123-25-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009208
InChI Key DKMROQRQHGEIOW-UHFFFAOYSA-N
Synonym diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
PubChem CID 31249
IUPAC Name diethyl butanedioate
SMILES CCOC(=O)CCC(=O)OCC

Specifications

Color Colorless
Boiling Point 196°C
Quantity 25 g
Formula Weight 174.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Succinate
EINECSNumber (2)-0848
RTECSNumber WM7400000
TSCA Yes