missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl succinate, 98%
CAS: 123-25-1 | C8H14O4 | 174.196 g/mol
$57.21 - $1,025.64
Chemical Identifiers
| CAS | 123-25-1 |
|---|---|
| Molecular Formula | C8H14O4 |
| Molecular Weight (g/mol) | 174.196 |
| MDL Number | MFCD00009208 |
| InChI Key | DKMROQRQHGEIOW-UHFFFAOYSA-N |
| Synonym | diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 |
| PubChem CID | 31249 |
| IUPAC Name | diethyl butanedioate |
| SMILES | CCOC(=O)CCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1577722
|
Thermo Scientific Chemicals
A1577722 |
100 g |
N/A
|
|
|||||
|
AAA1577736
|
Thermo Scientific Chemicals
A1577736 |
500 g |
N/A
|
|
|||||
|
AAA157770C
|
Thermo Scientific Chemicals
A157770C |
10,000 g |
N/A
|
|
|||||
Description
It is employed as emollients, masking agents, plasticisers and solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as emollients, masking agents, plasticisers and solvents.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
| 123-25-1 | |
| 174.196 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| 31249 | |
| CCOC(=O)CCC(=O)OCC |
| C8H14O4 | |
| MFCD00009208 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| diethyl butanedioate |
Specifications
| 123-25-1 | |
| 1.043 | |
| 90°C (194°F) | |
| 1.42 | |
| 100 g | |
| 14,8869 | |
| Slightly soluble in water. | |
| CCOC(=O)CCC(=O)OCC | |
| 174.196 | |
| 174.2 | |
| Diethyl succinate |
| -21°C | |
| 216°C to 218°C | |
| C8H14O4 | |
| MFCD00009208 | |
| 907645 | |
| diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377 | |
| DKMROQRQHGEIOW-UHFFFAOYSA-N | |
| diethyl butanedioate | |
| 31249 | |
| 98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 204-612-0
RTECSNumber : WM7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only