missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Suberate 98.0+%, TCI America™

Catalog No. S044925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S044925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S044925G Supplier TCI America Supplier No. S044925G

Chemical Identifiers

CAS 2050-23-9
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.30
MDL Number MFCD00009217
InChI Key PEUGOJXLBSIJQS-UHFFFAOYSA-N
Synonym diethyl suberate, ethyl suberate, octanedioic acid, diethyl ester, diethylsuberate, suberic acid diethyl ester, 1,8-diethyl octanedioate, acmc-1ce5z, ksc492k9j, octanedioic acid diethyl ester, octanedioic acid,1,8-diethyl ester
PubChem CID 16301
IUPAC Name 1,8-diethyl octanedioate
SMILES CCOC(=O)CCCCCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 283°C
Quantity 25 g
Formula Weight 230.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Suberate
EINECSNumber (2)-0879
TSCA No