missing translation for 'onlineSavingsMsg'
Learn More

Diethyl suberate, 99%

Catalog No. AAA1473536
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1473536 500 g
AAA1473518 50 g
AAA1473530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1473536 Supplier Thermo Scientific Chemicals Supplier No. A1473536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2050-23-9 | C12H22O4 | 230.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2050-23-9
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.30
MDL Number MFCD00009217
InChI Key PEUGOJXLBSIJQS-UHFFFAOYSA-N
Synonym diethyl suberate, ethyl suberate, octanedioic acid, diethyl ester, diethylsuberate, suberic acid diethyl ester, 1,8-diethyl octanedioate, acmc-1ce5z, ksc492k9j, octanedioic acid diethyl ester, octanedioic acid,1,8-diethyl ester
PubChem CID 16301
IUPAC Name diethyl octanedioate
SMILES CCOC(=O)CCCCCCC(=O)OCC

Specifications

Density 0.98
Boiling Point 282°C to 283°C
Flash Point >110°C (230°F)
Refractive Index 1.432
Linear Formula CH3CH2O2C(CH2)6CO2CH2CH3
Quantity 500 g
Beilstein 1785500
Formula Weight 230.3
Percent Purity 99%
Chemical Name or Material Diethyl suberate
EINECSNumber 218-084-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only