missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sec-Butylmalonate 95.0+%, TCI America™

Catalog No. B038425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B038425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B038425ML Supplier TCI America Supplier No. B038425ML
Only null left

Chemical Identifiers

CAS 83-27-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00015194
InChI Key MIIZSUOEOUHAIZ-UHFFFAOYSA-N
Synonym diethyl sec-butylmalonate, diethyl 2-sec-butyl malonate, sec-butylmalonic acid diethyl ester, diethyl 1-methylpropyl malonate, diethyl 1-methylpropylmalonate, sec-butylmalonic acid, diethyl ester, 1,3-diethyl 2-sec-butyl propanedioate, diethyl s-butylmalonate, diethylsec-butylmalonate, acmc-209prt
PubChem CID 78940
IUPAC Name diethyl 2-butan-2-ylpropanedioate
SMILES CCC(C)C(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 225°C
Quantity 25 mL
Formula Weight 216.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl sec-Butylmalonate
EINECSNumber (2)-0879
TSCA No