missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate Ribonuclease inhibitor. MP Biomedicals

Catalog No. icn15090250
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15090250 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15090250 Supplier MP Biomedicals Inc Supplier No. 0215090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name diethyl dicarbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Density 1.12 g/mL
Quantity 50 mL
Formula Weight 162.1
Chemical Name or Material Diethyl Pyrocarbonate, Ribonuclease inhibitor.