missing translation for 'onlineSavingsMsg'
Learn More

Pyrocarbonate diéthylique, MP Biomedicals

Catalog No. ICN15090225
Change view
Click to view available options
Quantité:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
ICN15090225 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15090225 Supplier MP Biomedicals Inc Supplier No. 0215090225

missing translation for 'validMainframeMsg'

Inhibiteur de la ribonucléase

Chemical Identifiers

CAS 1609-47-8
Formule moléculaire C6H10O5
Poids moléculaire (g/mol) 162.14
Numéro MDL MFCD00009106
Clé InChI FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonyme diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
Nom de l’IUPAC Carbonate d’éthyle ethoxycarbonyle
SOURIRES CCOC(=O)OC(=O)OCC

Specifications

Nom ou substance chimique Diethyl Pyrocarbonate
Densité Densité 1,12 g/ml, 1,1 sp. gr.
Quantité 25 mL
Poids de formule 162.14
Forme physique Liquid