missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, >97%, Ultrapure

Catalog No. p-7082786
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ14710AC 25 mL Plastic bottle
AAJ14710AE 100 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ14710AC Supplier Thermo Fisher Scientific Supplier No. J14710AC
Only null left

CAS: 1609-47-8 | C6H10O5 | 162.141 g/mol

  • A strong, nonspecific inhibitor of ribonuclease
  • Reacts with the N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Strong, nonspecific inhibitor of ribonuclease
  • Reacts with N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name ethoxycarbonyl ethyl carbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Melting Point 69°C
Density 1.101
Boiling Point 93°C to 94°C (18 mmHg)
Flash Point 69°C
Packaging Plastic bottle
Refractive Index 1.398
Quantity 25 mL
Beilstein 637031
Formula Weight 162.14
Percent Purity ≥97%
Grade Ultrapure
Physical Form Liquid
Chemical Name or Material Diethyl Pyrocarbonate
Show More Show Less
Hazard Category H227-H302-H315-H319-H332-H335
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 216-542-8
RTECSNumber LQ9350000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only