Learn More
Diethyl pyrocarbonate, 97%, pure
CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol
Supplier: Thermo Scientific Chemicals 170250250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Diethyl pyrocarbonate | |
| 1.1175g/mL | |
| 69°C | |
| Glass bottle | |
| 1.3960 to 1.4000 | |
| MFCD00009106 | |
| 03,IV,18 | |
| 153146 | |
| Solubility in water: slow decomposition. Other solubilities: very soluble in acetone,ethanol,ligroin | |
| CCOC(=O)OC(=O)OCC | |
| 162.14 | |
| 1.97 mPa.s (20°C) | |
| 162.14 | |
| Pure |
| 1609-47-8 | |
| 93.0°C to 94.0°C (18.0 mmHg) | |
| 0.97 | |
| C6H10O5 | |
| O(CO2C2H5)2 | |
| 25 g | |
| 1.1175 | |
| diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
| FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
| ethoxycarbonyl ethyl carbonate | |
| 3051 | |
| CHEBI:59051 | |
| 97% |
Chemical Identifiers
| 1609-47-8 | |
| 162.14 | |
| FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
| 3051 | |
| ethoxycarbonyl ethyl carbonate |
| C6H10O5 | |
| MFCD00009106 | |
| diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
| CHEBI:59051 | |
| CCOC(=O)OC(=O)OCC |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed, Combustible liquid, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Flammable liquids (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
EINECSNumber : 216-542-8
RUO – Research Use Only