missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrocarbonate, 97%, pure

Catalog No. AC170250050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170250050 5 g Glass bottle
AC170250250 25 g Glass bottle
AC170251000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC170250050 Supplier Thermo Scientific Chemicals Supplier No. 170250050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name ethoxycarbonyl ethyl carbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Density 1.1175g/mL
Boiling Point 93.0°C to 94.0°C (18.0 mmHg)
Flash Point 69°C
Assay Percent Range 0.97
Packaging Glass bottle
Refractive Index 1.3960 to 1.4000
Linear Formula O(CO2C2H5)2
Quantity 5 g
Beilstein 03,IV,18
Merck Index 153146
Specific Gravity 1.1175
Solubility Information Solubility in water: slow decomposition. Other solubilities: very soluble in acetone,ethanol,ligroin
Viscosity 1.97 mPa.s (20°C)
Formula Weight 162.14
Percent Purity 97%
Grade Pure
Chemical Name or Material Diethyl pyrocarbonate
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Flammable liquids (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, Harmful if swallowed, Combustible liquid, May cause respiratory irritation, Causes skin irritation
EINECSNumber 216-542-8

RUO – Research Use Only