missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Propylmalonate 98.0+%, TCI America™

Catalog No. D47085ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47085ML 5 mL
D470825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47085ML Supplier TCI America Supplier No. D47085ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2163-48-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009168
InChI Key GRRSDGHTSMJICM-UHFFFAOYSA-N
Synonym diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate
PubChem CID 16552
IUPAC Name diethyl 2-propylpropanedioate
SMILES CCCC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 222°C
Quantity 5 mL
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Propylmalonate
EINECSNumber (2)-0861
TSCA No