missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phosphoramidate 98.0+%, TCI America™

Catalog No. D23615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23615G 5 g
D236125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23615G Supplier TCI America Supplier No. D23615G
Only null left

Chemical Identifiers

CAS 1068-21-9
Molecular Formula C4H12NO3P
Molecular Weight (g/mol) 153.118
MDL Number MFCD00015676
InChI Key MCQILDHFZKTBOD-UHFFFAOYSA-N
Synonym Diethyl Amidophosphate, Diethyl Phosphoryl Amide
PubChem CID 70605
IUPAC Name [amino(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(N)OCC

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 153.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Phosphoramidate
EINECSNumber (2)-1992
TSCA No