missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog No. p-109785
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114502500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC114502500 Supplier Thermo Scientific Chemicals Supplier No. 114502500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762-04-9
MDL Number MFCD00044573
InChI Key LXCYSACZTOKNNS-UHFFFAOYSA-N
Synonym diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
PubChem CID 6327654
SMILES CCO[P+](=O)OCC

Specifications

Melting Point -70°C
Color Colorless
Density 1.0800g/mL
Boiling Point 60°C (6.0 Mbar)
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H11O3P
Refractive Index 1.4070 to 1.409
Linear Formula (C2H5O)2P(O)H
Quantity 250 g
Beilstein 01, 330
Fieser 01,251; 02,132; 12,187
Specific Gravity 1.08
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in common organic solvents
IUPAC Name diethoxy(oxo)phosphanium
Molecular Weight (g/mol) 138.1
Viscosity 103 mm2/s (37°C)
Formula Weight 138.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl phosphite, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 212-091-6
RTECSNumber TG7875000
TSCA TSCA

RUO – Research Use Only