missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphite, 96%, Thermo Scientific Chemicals

Catalog No. p-7052191
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4380130 250 g
AA43801A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4380130 Supplier Thermo Scientific Chemicals Supplier No. 04380130

Intermediate for preparation of phosphonate esters

Diethyl phosphite used as intermediate for extraction agent and phosphoric acid esters. It is also used as solvent and acid acceptor in the synthesis of polypeptide. It is also used in the synthetic reaction. It is a precursor of phosphorous which is used for the synthesis of various organophosphorus compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762-04-9
Molecular Formula C4H10O3P+
Molecular Weight (g/mol) 137.095
MDL Number MFCD00044573
InChI Key LXCYSACZTOKNNS-UHFFFAOYSA-N
Synonym diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
PubChem CID 6327654
IUPAC Name diethoxy(oxo)phosphanium
SMILES CCO[P+](=O)OCC

Specifications

Melting Point -70°C
Density 1.070 g/mL
Boiling Point 50°C to 51°C (2 mmHg)
Flash Point 74°C (165°F)
Odor Weak
Quantity 250 g
Sensitivity Moisture sensitive
Solubility Information Miscible in water.
Formula Weight 138.1
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl phosphite
Show More Show Less
Hazard Category H227-H317-H318
Hazard Statement GHS H Statement
H318-H317-H227-H315
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
EINECSNumber 212-091-6
RTECSNumber TG7875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only