missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphate, 96%

Catalog No. AC407420050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407420050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407420050 Supplier Thermo Scientific Chemicals Supplier No. 407420050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-02-7 | C4H11O4P | 154.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-02-7 , 7664-38-2
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.1
InChI Key UCQFCFPECQILOL-UHFFFAOYSA-N
Synonym diethyl phosphate, diethyl acid phosphate, phosphoric acid, diethyl ester, diethyl phosphoric acid, diethylphosphate, diethylphosphoric acid, ethyl phosphate, di, o,o-diethyl hydrogen phosphate, ethyl phosphate, eto 2 ho po, o,o-diethylphosphoric acid
PubChem CID 654
ChEBI CHEBI:27708
IUPAC Name diethyl hydrogen phosphate
SMILES CCOP(=O)(O)OCC

Specifications

Melting Point 6.0°C
Density 1.2900g/mL
Boiling Point 203.0°C
Flash Point 91°C
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula (C2H5O)2PO2H
Quantity 5 g
Specific Gravity 1.29
Solubility Information Solubility in water: slightly soluble.
Formula Weight 154.1
Percent Purity 96%
Chemical Name or Material Diethyl phosphate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 209-912-5

RUO – Research Use Only