Learn More
Diethyl phenylthiomethylphosphonate, 96%
CAS: 38066-16-9 | C11H17O3PS | 260.288 g/mol
Supplier: Thermo Scientific Chemicals A1366018
Description
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| Diethyl phenylthiomethylphosphonate | |
| 1.174 | |
| C11H17O3PS | |
| MFCD00015130 | |
| 1246215 | |
| Difficult to mix. | |
| CCOP(=O)(CSC1=CC=CC=C1)OCC | |
| 260.288 | |
| 260.29 |
| 38066-16-9 | |
| 146°C to 148°C (0.5 mmHg) | |
| 1.534 | |
| 50 g | |
| diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate # | |
| FBUXEPJJFVDUFE-UHFFFAOYSA-N | |
| diethoxyphosphorylmethylsulfanylbenzene | |
| 99227 | |
| 96% |
Chemical Identifiers
| 38066-16-9 | |
| 260.288 | |
| FBUXEPJJFVDUFE-UHFFFAOYSA-N | |
| 99227 | |
| CCOP(=O)(CSC1=CC=CC=C1)OCC |
| C11H17O3PS | |
| MFCD00015130 | |
| diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate # | |
| diethoxyphosphorylmethylsulfanylbenzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 253-770-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only