missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylthiomethylphosphonate, 96%

Catalog No. AAA1366009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366009 10 g
AAA1366018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1366009 Supplier Thermo Scientific Chemicals Supplier No. A1366009

CAS: 38066-16-9 | C11H17O3PS | 260.288 g/mol

Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 38066-16-9
Molecular Formula C11H17O3PS
Molecular Weight (g/mol) 260.288
MDL Number MFCD00015130
InChI Key FBUXEPJJFVDUFE-UHFFFAOYSA-N
Synonym diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate #
PubChem CID 99227
IUPAC Name diethoxyphosphorylmethylsulfanylbenzene
SMILES CCOP(=O)(CSC1=CC=CC=C1)OCC

Specifications

Density 1.174
Boiling Point 146°C to 148°C (0.5 mmHg)
Refractive Index 1.534
Quantity 10 g
Beilstein 1246215
Solubility Information Difficult to mix.
Formula Weight 260.29
Percent Purity 96%
Chemical Name or Material Diethyl phenylthiomethylphosphonate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-770-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only