missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phenylphosphonate 96.0+%, TCI America™

Catalog No. P03355ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03355ML 5 mL
P033525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03355ML Supplier TCI America Supplier No. P03355ML

Chemical Identifiers

CAS 1754-49-0
Molecular Formula C10H15O3P
Molecular Weight (g/mol) 214.20
MDL Number MFCD00015136
InChI Key VZEGPPPCKHRYGO-UHFFFAOYSA-N
Synonym diethyl phenylphosphonate, diethyl benzenephosphonate, phenylphosphonic acid diethyl ester, phosphonic acid, phenyl-, diethyl ester, benzenephosphonic acid, diethyl ester, diethoxyphosphoryl-benzene, acmc-1c01k, phenylphosphonic acid diethyl, wln: 2vopo&r&ov2, benzenephosphonic acid diethyl ester
PubChem CID 15642
IUPAC Name diethyl phenylphosphonate
SMILES CCOP(=O)(OCC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 267°C
Quantity 5 mL
Formula Weight 214.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Phenylphosphonate
EINECSNumber (3)-2521
RTECSNumber TA0377000
TSCA No