missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Phenylmalonate 97.0+%, TCI America™
$36.57 - $186.74
Chemical Identifiers
| CAS | 83-13-6 |
|---|---|
| Molecular Formula | C13H16O4 |
| Molecular Weight (g/mol) | 236.267 |
| MDL Number | MFCD00009144 |
| InChI Key | FGYDHYCFHBSNPE-UHFFFAOYSA-N |
| Synonym | diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate |
| PubChem CID | 66514 |
| IUPAC Name | diethyl 2-phenylpropanedioate |
| SMILES | CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P031125ML
|
TCI America
P031125ML |
25 mL |
Each for $36.57
|
|
|||||
|
P0311500ML
|
TCI America
P0311500ML |
500 mL |
Each for $186.74
|
|
|||||
Chemical Identifiers
| 83-13-6 | |
| 236.267 | |
| FGYDHYCFHBSNPE-UHFFFAOYSA-N | |
| 66514 | |
| CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC |
| C13H16O4 | |
| MFCD00009144 | |
| diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate | |
| diethyl 2-phenylpropanedioate |
Specifications
| 83-13-6 | |
| 285°C | |
| MFCD00009144 | |
| diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate | |
| CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC | |
| 236.267 | |
| 236.27 | |
| Liquid |
| Yellow | |
| C13H16O4 | |
| 25 mL | |
| FGYDHYCFHBSNPE-UHFFFAOYSA-N | |
| diethyl 2-phenylpropanedioate | |
| 66514 | |
| ≥97.0% (GC) | |
| Diethyl Phenylmalonate |
Safety and Handling
EINECSNumber : (9)-1175
TSCA : Yes