missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phenylmalonate 97.0+%, TCI America™

Catalog No. P031125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P031125ML 25 mL
P0311500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P031125ML Supplier TCI America Supplier No. P031125ML

Chemical Identifiers

CAS 83-13-6
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.267
MDL Number MFCD00009144
InChI Key FGYDHYCFHBSNPE-UHFFFAOYSA-N
Synonym diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
PubChem CID 66514
IUPAC Name diethyl 2-phenylpropanedioate
SMILES CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC

Specifications

Color Yellow
Boiling Point 285°C
Quantity 25 mL
Formula Weight 236.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Phenylmalonate
EINECSNumber (9)-1175
TSCA Yes