missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

Catalog No. p-7024958
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
Catalog No. Quantity
AAB2023430 250 g
1 options

Catalog No. AAB2023430

Supplier: Thermo Scientific Chemicals B2023430

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-13-6
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.267
MDL Number MFCD00009144
InChI Key FGYDHYCFHBSNPE-UHFFFAOYSA-N
Synonym diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
PubChem CID 66514
IUPAC Name diethyl 2-phenylpropanedioate
SMILES CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 15°C to 16°C
Density 1.096
Boiling Point 170°C to 172°C (14 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.491
Quantity 250 g
Beilstein 614465
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material Diethyl phenylmalonate
EINECSNumber 201-456-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only