missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenyl orthoformate, 97%

Catalog No. AAB2149809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2149809 10 g
AAB2149818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2149809 Supplier Thermo Scientific Chemicals Supplier No. B2149809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14444-77-0 | C11H16O3 | 196.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14444-77-0
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00009228
InChI Key QTURWMMVIIBRRP-UHFFFAOYSA-N
Synonym diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester
PubChem CID 84440
IUPAC Name diethoxymethoxybenzene
SMILES CCOC(OCC)OC1=CC=CC=C1

Specifications

Melting Point -26°C
Density 1.015
Boiling Point 103°C to 104°C (10 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.4795
Quantity 10 g
Beilstein 1872634
Sensitivity Moisture sensitive
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Diethyl phenyl orthoformate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 238-421-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only