missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Oxalate 98.0+%, TCI America™

Catalog No. O007825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O007825G 25 g
O0078500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O007825G Supplier TCI America Supplier No. O007825G

Chemical Identifiers

CAS 95-92-1
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00009119
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
Synonym ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove
PubChem CID 7268
IUPAC Name diethyl oxalate
SMILES CCOC(=O)C(=O)OCC

Specifications

Melting Point -41°C
Color Colorless
Boiling Point 185°C
Quantity 25 g
UN Number 2525
Formula Weight 146.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Oxalate
EINECSNumber (2)-0924
RTECSNumber RO2800000
TSCA Yes