Learn More
Diethyl oxalate, 99%
CAS: 95-92-1 | C6H10O4 | 146.142 g/mol
Supplier: Thermo Scientific Chemicals A144980C
Description
Diethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.
Solubility
Miscible with alcohols, ether and other common organic solvents.
Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases. Moisture sensitive.
Specifications
| Diethyl oxalate | |
| -41°C | |
| 184°C to 186°C | |
| C6H10O4 | |
| MFCD00009119 | |
| UN2525 | |
| Moisture sensitive | |
| ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove | |
| WYACBZDAHNBPPB-UHFFFAOYSA-N | |
| diethyl oxalate | |
| 7268 | |
| 99% |
| 95-92-1 | |
| 1.077 | |
| 75°C (167°F) | |
| 1.41 | |
| 10,000 g | |
| 606350 | |
| 14,3125 | |
| Miscible with alcohols,ether and other common organic solvents. | |
| CCOC(=O)C(=O)OCC | |
| 146.142 | |
| 146.14 |
Chemical Identifiers
| 95-92-1 | |
| 146.142 | |
| WYACBZDAHNBPPB-UHFFFAOYSA-N | |
| 7268 | |
| CCOC(=O)C(=O)OCC |
| C6H10O4 | |
| MFCD00009119 | |
| ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove | |
| diethyl oxalate |
Safety and Handling
GHS H Statement
H302-H319-H227
Harmful if swallowed.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H227-H302-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ETHYL OXALATE
EINECSNumber : 202-464-1
RTECSNumber : RO2800000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only