missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Oxalacetate 95.0+%, TCI America™

Catalog No. p-7134172
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00735G 5 g
O007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00735G Supplier TCI America Supplier No. O00735G
Only null left

Chemical Identifiers

CAS 108-56-5
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.179
MDL Number MFCD00059229
InChI Key JDXYSCUOABNLIR-UHFFFAOYSA-N
Synonym Ethyl Oxalacetate, Oxalacetic Acid Diethyl Ester
PubChem CID 66951
ChEBI CHEBI:86608
IUPAC Name diethyl 2-oxobutanedioate
SMILES CCOC(=O)CC(=O)C(=O)OCC

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 188.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Oxalacetate
EINECSNumber (2)-1344
TSCA No
Recommended Storage Freezer