missing translation for 'onlineSavingsMsg'
Learn More

Diethyl oxalacetate sodium salt, 95%, pract.

CAS: 40876-98-0 | C8H11NaO5 | 210.16 g/mol

$99.16 - $99.16

Chemical Identifiers

CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.16
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC407371000
SDS View Documents
Thermo Scientific Chemicals
407371000
100 g
Each for $99.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40876-98-0
210.16
UJZUICGIJODKOS-UHFFFAOYSA-M
44134875
CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
C8H11NaO5
MFCD00035571
diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate

Specifications

40876-98-0
Authentic
Plastic bottle
C2H5OCOCH=C(ONa)COOC2H5
100 g
diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
210.16
210.16
Practical
Diethyl oxalacetate sodium salt
Beige to Orange
95%
C8H11NaO5
MFCD00035571
14, 3836
UJZUICGIJODKOS-UHFFFAOYSA-M
sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
44134875
95%
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 255-122-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only