missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Oxalacetate Sodium Salt 95.0+%, TCI America™
$1183.98 - $1183.98
Chemical Identifiers
| CAS | 40876-98-0 |
|---|---|
| Molecular Formula | C8H11NaO5 |
| Molecular Weight (g/mol) | 210.161 |
| MDL Number | MFCD00035571 |
| InChI Key | UJZUICGIJODKOS-UHFFFAOYSA-M |
| Synonym | diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n |
| PubChem CID | 44134875 |
| IUPAC Name | sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate |
| SMILES | CCOC(=O)C=C(C(=O)OCC)[O-].[Na+] |
Chemical Identifiers
| 40876-98-0 | |
| 210.161 | |
| UJZUICGIJODKOS-UHFFFAOYSA-M | |
| 44134875 | |
| CCOC(=O)C=C(C(=O)OCC)[O-].[Na+] |
| C8H11NaO5 | |
| MFCD00035571 | |
| diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n | |
| sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate |
Specifications
| 40876-98-0 | |
| MFCD00035571 | |
| diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n | |
| CCOC(=O)C=C(C(=O)OCC)[O-].[Na+] | |
| 210.161 | |
| 210.16 | |
| Crystalline Powder |
| C8H11NaO5 | |
| 500 g | |
| UJZUICGIJODKOS-UHFFFAOYSA-M | |
| sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate | |
| 44134875 | |
| ≥95.0% (T) | |
| Diethyl Oxalacetate Sodium Salt |
Safety and Handling
EINECSNumber : (2)-2624&(2)-2735
RTECSNumber : EK0115500
TSCA : Yes