missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Oxalacetate Sodium Salt 95.0+%, TCI America™

Catalog No. O0074500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0074500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0074500G Supplier TCI America Supplier No. O0074500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.161
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]

Specifications

Quantity 500 g
Formula Weight 210.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Oxalacetate Sodium Salt
EINECSNumber (2)-2624&(2)-2735
RTECSNumber EK0115500
TSCA Yes