missing translation for 'onlineSavingsMsg'
Learn More

Diethyl N-(tert-Butoxycarbonyl)phosphoramidate 98.0+%, TCI America™

Catalog No. B17345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17345G Supplier TCI America Supplier No. B17345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85232-02-6
Molecular Formula C9H20NO5P
Molecular Weight (g/mol) 253.235
MDL Number MFCD00015667
InChI Key MJJRHRHTDZXUKY-UHFFFAOYSA-N
Synonym N-Boc-phosphoramidic Acid Diethyl Ester, N-(tert-Butoxycarbonyl)phosphoramidic Acid Diethyl Ester, Diethyl N-Boc-phosphoramidate
PubChem CID 9837914
IUPAC Name tert-butyl N-diethoxyphosphorylcarbamate
SMILES CCOP(=O)(NC(=O)OC(C)(C)C)OCC

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 5 g
Formula Weight 253.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl N-(tert-Butoxycarbonyl)phosphoramidate