missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate 97.0+%, TCI America™
Supplier: TCI America D37085G
Specifications
| Diethyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate | |
| Yellow | |
| C8H18NO5P | |
| 5 g | |
| WYLRYBDGIILFIR-UHFFFAOYSA-N | |
| 2-diethoxyphosphoryl-N-methoxy-N-methylacetamide | |
| 2773719 | |
| ≥97.0% (GC) |
| 124931-12-0 | |
| 124°C | |
| MFCD00134233 | |
| (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diethyl Ester | |
| CCOP(=O)(CC(=O)N(C)OC)OCC | |
| 239.208 | |
| 239.21 | |
| Liquid |
Chemical Identifiers
| 124931-12-0 | |
| 239.208 | |
| WYLRYBDGIILFIR-UHFFFAOYSA-N | |
| 2773719 | |
| CCOP(=O)(CC(=O)N(C)OC)OCC |
| C8H18NO5P | |
| MFCD00134233 | |
| (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diethyl Ester | |
| 2-diethoxyphosphoryl-N-methoxy-N-methylacetamide |
Safety and Handling
TSCA : No