missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate 97.0+%, TCI America™

Catalog No. D37081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37081G 1 g
D37085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37081G Supplier TCI America Supplier No. D37081G

Chemical Identifiers

CAS 124931-12-0
Molecular Formula C8H18NO5P
Molecular Weight (g/mol) 239.208
MDL Number MFCD00134233
InChI Key WYLRYBDGIILFIR-UHFFFAOYSA-N
Synonym (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diethyl Ester
PubChem CID 2773719
IUPAC Name 2-diethoxyphosphoryl-N-methoxy-N-methylacetamide
SMILES CCOP(=O)(CC(=O)N(C)OC)OCC

Specifications

Color Yellow
Boiling Point 124°C
Quantity 1 g
Formula Weight 239.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (N-Methoxy-N-methylcarbamoylmethyl)phosphonate